Learn More
Medchemexpress LLC Alpha-Lactose | 14641-93-1 | 1 G
SDP
Catalog No. 5000421469
Encompass_Preferred
Alpha-Lactose (alpha-D-Lactose) is the main sugar found in milk. Lactose exists in two anomeric forms, alpha and beta, with the alpha form typically crystallizing as a monohydrate. It is an important sugar due to its abundance in the milk of humans and domestic animals, serving as a basic nutrient and a primary substrate in fermentation processes for products like yogurt and kefir. This product is for research use only and not sold to patients.
  • Main sugar found in milk
  • Exists in two anomeric forms, alpha and beta
  • Alpha form typically crystallizes as a monohydrate
  • Important nutrient in milk
  • Primary substrate in fermentation processes
  • For research use only
Catalog No. 50-004-21469 Supplier Medchemexpress LLC Supplier No. HYN25141G
Alpha-Lactose (alpha-D-Lactose) is the main sugar found in milk. Lactose exists in two anomeric forms, alpha and beta, with the alpha form typically crystallizing as a monohydrate. It is an important sugar due to its abundance in the milk of humans and domestic animals, serving as a basic nutrient and a primary substrate in fermentation processes for products like yogurt and kefir. This product is for research use only and not sold to patients.
  • Main sugar found in milk
  • Exists in two anomeric forms, alpha and beta
  • Alpha form typically crystallizes as a monohydrate
  • Important nutrient in milk
  • Primary substrate in fermentation processes
  • For research use only