Learn More
Selleck Chemical LLC ()-a-Lipoic acid S3998-25mg
SDP
Catalog No. 503108687
Encompass_Preferred
( )- -Lipoic acid ((R)-( )- -Lipoic acid -Lipoic acid Alpha-Lipoic acid) a physiological form of thioctic acid is a strong antioxidant that relieves diabetic neuropathic symptoms It shows superior antioxidative effects to its racemate R( )- -lipoic acid is an essential cofactor of mitochondrial enzyme complexes R( )- -lipoic acid inhibits NF- B-dependent HIV-1 LTR activation
Catalog No. 50-310-8687 Supplier Selleck Chemical LLC Supplier No. S399825MG
( )- -Lipoic acid ((R)-( )- -Lipoic acid -Lipoic acid Alpha-Lipoic acid) a physiological form of thioctic acid is a strong antioxidant that relieves diabetic neuropathic symptoms It shows superior antioxidative effects to its racemate R( )- -lipoic acid is an essential cofactor of mitochondrial enzyme complexes R( )- -lipoic acid inhibits NF- B-dependent HIV-1 LTR activation