Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-Alpha-Lipoic Acid

Catalog No. 6232025GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-320-25GF 25 g
62-320-5GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 62-320-25GF Supplier Sigma Aldrich Supplier No. 6232025GF
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(+/-)-Alpha-Lipoic Acid

Specifications

CAS 1077-28-7
Melting Point 60°C to 62°C
Molecular Formula C8H14O2S2
Linear Formula C8H14O2S2
MDL Number MFCD00005474
Quantity 25 g
Synonym (+/-)-1,2-Dithiolane-3-pentanoic acid; 6,8-Dithiooctanoic acid; DL-alpha-Lipoic acid; DL-6,8-Thioctic acid; Lip(S2)
Molecular Weight (g/mol) 206.33 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.