Learn More
Medchemexpress LLC Alpha-lipoic acid choline ester chloride | 1808266-58-1 | 99.7% | 327.93 | C13H26ClNO2S2 | 5 MG
SDP
Catalog No. 5000221825
Encompass_Preferred
Alpha-lipoic acid choline ester chloride is a choline ester derivative of alpha-lipoic acid provided for laboratory research. It can reduce protein disulfides and has been studied for improving lens elasticity and other presbyopia-related effects. The compound is supplied as a solid and as prepared DMSO solutions; store sealed at 4°C and handle per standard laboratory safety procedures.
  • Choline ester derivative of alpha-lipoic acid
  • Reduces protein disulfides
  • Enhances lens elasticity in preclinical studies
  • Available as solid and 10 mM DMSO solutions
  • High purity (99.7%)
  • Molecular formula C13H26ClNO2S2; molecular weight 327.93
Catalog No. 50-002-21825 Supplier Medchemexpress LLC Supplier No. HY1346335MG
Alpha-lipoic acid choline ester chloride is a choline ester derivative of alpha-lipoic acid provided for laboratory research. It can reduce protein disulfides and has been studied for improving lens elasticity and other presbyopia-related effects. The compound is supplied as a solid and as prepared DMSO solutions; store sealed at 4°C and handle per standard laboratory safety procedures.
  • Choline ester derivative of alpha-lipoic acid
  • Reduces protein disulfides
  • Enhances lens elasticity in preclinical studies
  • Available as solid and 10 mM DMSO solutions
  • High purity (99.7%)
  • Molecular formula C13H26ClNO2S2; molecular weight 327.93