Learn More
Medchemexpress LLC Alpha-lipoic acid choline ester chloride | 1808266-58-1 | 99.7% | C13H26ClNO2S2 | 10MG
SDP
Catalog No. 5000260705
Encompass_Preferred
Alpha-lipoic acid choline ester chloride is a chemically modified derivative of alpha-lipoic acid provided as a research-grade solid for biochemical and pharmacological studies. It has molecular formula C13H26ClNO2S2, molecular weight 327.93 g/mol, and high purity suitable for analytical and in vitro applications.
  • High purity (99.7%) suitable for analytical and in vitro studies.
  • Available in small milligram quantities for screening and assay work.
  • Off-white to light yellow solid, easy to handle and store.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Useful as a reagent in studies of lipoic acid derivatives and choline conjugates.
Catalog No. 50-002-60705 Supplier Medchemexpress LLC Supplier No. HY13463310MG
Alpha-lipoic acid choline ester chloride is a chemically modified derivative of alpha-lipoic acid provided as a research-grade solid for biochemical and pharmacological studies. It has molecular formula C13H26ClNO2S2, molecular weight 327.93 g/mol, and high purity suitable for analytical and in vitro applications.
  • High purity (99.7%) suitable for analytical and in vitro studies.
  • Available in small milligram quantities for screening and assay work.
  • Off-white to light yellow solid, easy to handle and store.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Useful as a reagent in studies of lipoic acid derivatives and choline conjugates.