Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Lipoic Acid Dietary Supplement, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18600435 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-435 1 kg Amber Glass Bottle
18-600-432 5 g Amber Glass Bottle
18-600-433 25 g Amber Glass Bottle
18-600-434 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-435 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L15061KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

L1506, 1077-28-7, C8H14O2S2

Spectrum™ Chemical Alpha Lipoic Acid, USP Dietary Supplement is used for diabetes and nerve-related symptoms of diabetes including burning, pain, and numbness in the legs and arms. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

CAS Max % 100%
Boiling Point 160°C to 165 °C (320°F to 329 °F)
Ignition Residue 0.1%
Loss on Drying 0.2%
Melting Range 60-62°C
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation -1.0° to +1.0°
Percent Purity 99 to 101%
Grade USP Dietary Supplement
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Alpha Lipoic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.