Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-Lipoic acid-NHS | 40846-94-4 | 98.0% | C12H17NO4S2 | 100 MG
SDP

Catalog No. 5000353207
Encompass_Preferred

Alpha-Lipoic acid-NHS is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are designed to contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • Alkyl/ether based PROTAC linker
  • Used in synthesis of PROTACs
  • Solid appearance
  • White to yellow color
  • Molecular weight of 303.40
  • Purity of 98.0%

Catalog No. 50-003-53207 Supplier Medchemexpress LLC Supplier No. HY141336100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-Lipoic acid-NHS is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are designed to contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.

  • Alkyl/ether based PROTAC linker
  • Used in synthesis of PROTACs
  • Solid appearance
  • White to yellow color
  • Molecular weight of 303.40
  • Purity of 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.