Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-lipoic acid-NHS | 40846-94-4 | 98.0% | C12H17NO4S2 | 250 MG
SDP

Catalog No. 5000352782
Encompass_Preferred

Alpha-lipoic acid-NHS is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This compound has a molecular weight of 303.40 and is a white to yellow solid, intended for research use only.

  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Molecular weight of 303.40
  • Appears as a white to yellow solid
  • Shipping at room temperature (continental US)
  • Storage at -20°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in ethanol: 2 mg/mL (6.59 mM) with ultrasonic and warming to 60°C

Catalog No. 50-003-52782 Supplier Medchemexpress LLC Supplier No. 250MGHY141336
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-lipoic acid-NHS is an alkyl/ether-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs feature two ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system. This compound has a molecular weight of 303.40 and is a white to yellow solid, intended for research use only.

  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Molecular weight of 303.40
  • Appears as a white to yellow solid
  • Shipping at room temperature (continental US)
  • Storage at -20°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Solubility in ethanol: 2 mg/mL (6.59 mM) with ultrasonic and warming to 60°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.