Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-Lipoic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620965 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-965 1 g
18-620-966 5 g
18-620-967 25 g
18-620-968 100 g
18-620-969 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-965 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L15151GM
Add to Cart
Add to Cart

Synonym: DL-Thioctic Acid, Thioctic Acid, 5-(1,2-Dithiolan-3-yl)pentanoic Acid

Alpha-Lipoic Acid is also known as alpha lipoic acid or thioctic acid. It is an organosulfur compound that comes from octanoic acid. It is made most often in animals and is crucial for aerobic metabolism. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 1077-28-7

Specifications

Boiling Point 160°C to 165 °C (320°F to 329 °F)
Quantity 1 g
Molecular Weight (g/mol) 206.33
Percent Purity 98%
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material alpha-Lipoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.