Learn More
Enzo Life Sciences alpha-Mannosyl Ceramide (1mg)
SDP
Supplier:  Enzo Life Sciences ALX306031M001
Encompass_Preferred
Potent stimulator of natural killer T (NKT) cells expressing the invariant Vα19-Jα26 T cell receptor α chain (Vα19 NKT cells). Alternative name: alpha-ManCer. Purity: ≥95% (TLC and NMR). Solubility: Soluble in pyridine or warm DMSO (4mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8665