Learn More
CPC Scientific H-Trp-His-Trp-Leu-Gln-Leu-Lys-Pro-Gly-Gln-Pro-Met-Tyr-OH (trifluoroacetate salt) 5MG
SDP
Catalog No. 502110395
Encompass
This tridecapeptide, an alpha-factor pheromone of Saccharomyces cerevisiae, induces conjugation in yeast by binding to Ste2p. Its cognate GPCR activates a G protein signal pathway that highly conserves with mammalian signaling pathways. ONE-LETTER SEQUENCE: WHWLQLKPGQPMYMOLECULAR FORMULA: C82H114N20O17S1MOLECULAR WEIGHT:1684STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [59401-28-4], RESEARCH AREA: HormonalREFERENCES: Lee, BK. et al. J. Biol. Chem. 276, 37950 (2001); Mayrhofer, S. and S. Pöggeler Eukaryot. Cell 4, 661 (2005).
Catalog No. 50-211-0395 Supplier CPC Scientific Supplier No. SIGN001B
This tridecapeptide, an alpha-factor pheromone of Saccharomyces cerevisiae, induces conjugation in yeast by binding to Ste2p. Its cognate GPCR activates a G protein signal pathway that highly conserves with mammalian signaling pathways. ONE-LETTER SEQUENCE: WHWLQLKPGQPMYMOLECULAR FORMULA: C82H114N20O17S1MOLECULAR WEIGHT:1684STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [59401-28-4], RESEARCH AREA: HormonalREFERENCES: Lee, BK. et al. J. Biol. Chem. 276, 37950 (2001); Mayrhofer, S. and S. Pöggeler Eukaryot. Cell 4, 661 (2005).