Learn More
Medchemexpress LLC Alpha-Melanocyte-Stimulating Hormone TFA | 171869-93-5 | 98.5% | 1 MG
SDP
Catalog No. 5000425519
Encompass_Preferred
α-MSH TFA (α-Melanocyte-Stimulating Hormone TFA) is an endogenous neuropeptide and a melanocortin receptor 4 (MC4R) agonist. It exhibits anti-inflammatory and antipyretic activities and is a post-translational derivative of pro-opiomelanocortin (POMC).
  • Endogenous neuropeptide
  • MC4R agonist with anti-inflammatory and antipyretic activities
  • Post-translational derivative of pro-opiomelanocortin (POMC)
  • Modulates CNS inflammation by acting directly on melanocortin receptors in glial cells
  • Modulates NFκB activation
  • Inhibits translocation of transcription factor κB to the nucleus
Catalog No. 50-004-25519 Supplier Medchemexpress LLC Supplier No. HYP0252A1MG
α-MSH TFA (α-Melanocyte-Stimulating Hormone TFA) is an endogenous neuropeptide and a melanocortin receptor 4 (MC4R) agonist. It exhibits anti-inflammatory and antipyretic activities and is a post-translational derivative of pro-opiomelanocortin (POMC).
  • Endogenous neuropeptide
  • MC4R agonist with anti-inflammatory and antipyretic activities
  • Post-translational derivative of pro-opiomelanocortin (POMC)
  • Modulates CNS inflammation by acting directly on melanocortin receptors in glial cells
  • Modulates NFκB activation
  • Inhibits translocation of transcription factor κB to the nucleus