Learn More
STEMCELL Technologies Alpha MEM without Nucleosides, Size: 500 mL
SDP
Catalog No. NC1083740
Encompass
Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition. MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium.
Catalog No. NC1083740 Supplier STEMCELL Technologies Supplier No. 36453
Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications. Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition. MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium.