Learn More
Medchemexpress LLC Alpha-methyl-4-propylphenylacetic acid | 3585-47-5 | 98.0% | 192.25 g/mol | C12H16O2 | 5 MG
SDP
Catalog No. 5000262310
Encompass_Preferred
Ibuprofen impurity 1 (alpha-methyl-4-propylphenylacetic acid) is a characterized impurity of ibuprofen provided for research and analytical use. It is a solid organic acid with formula C12H16O2 and molecular weight 192.25 g/mol, supplied at high reported purity for impurity profiling, method development, and assay control in medicinal chemistry and COX inhibition studies.
  • High reported purity suitable for analytical applications.
  • Solid form for straightforward handling and storage.
  • Appropriate for impurity profiling and analytical method development.
  • Molecular formula C12H16O2 and molar mass 192.25 g/mol.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-62310 Supplier Medchemexpress LLC Supplier No. HY1312585MG
Ibuprofen impurity 1 (alpha-methyl-4-propylphenylacetic acid) is a characterized impurity of ibuprofen provided for research and analytical use. It is a solid organic acid with formula C12H16O2 and molecular weight 192.25 g/mol, supplied at high reported purity for impurity profiling, method development, and assay control in medicinal chemistry and COX inhibition studies.
  • High reported purity suitable for analytical applications.
  • Solid form for straightforward handling and storage.
  • Appropriate for impurity profiling and analytical method development.
  • Molecular formula C12H16O2 and molar mass 192.25 g/mol.
  • Available in small research pack sizes for laboratory use.