Learn More
Medchemexpress LLC Alpha-methyl-5-(2-thienylmethoxy)-1H-indole-3-ethanamine hydrochloride | 160521-72-2 | MFCD01321066 | 99.8% | 322.85 g/mol | C16H19ClN2OS | 100 MG
SDP
Catalog No. 5000211830
Encompass_Preferred
BW-723C86 is an orally active, selective 5-HT2B receptor agonist used in research to probe serotonergic signaling and related physiological responses in preclinical models.
  • Selective 5-HT2B receptor agonist with documented in vivo activity.
  • High purity suitable for research use (reported 99.8%).
  • Solid form, white to light brown, with solvent stability data.
  • High solubility in DMSO (≈100 mg/mL) for in vitro work.
  • In vivo formulation protocols available to achieve ≥5 mg/mL.
  • Multiple package sizes available for small-scale and bulk needs.
Catalog No. 50-002-11830 Supplier Medchemexpress LLC Supplier No. HY101369100MG
BW-723C86 is an orally active, selective 5-HT2B receptor agonist used in research to probe serotonergic signaling and related physiological responses in preclinical models.
  • Selective 5-HT2B receptor agonist with documented in vivo activity.
  • High purity suitable for research use (reported 99.8%).
  • Solid form, white to light brown, with solvent stability data.
  • High solubility in DMSO (≈100 mg/mL) for in vitro work.
  • In vivo formulation protocols available to achieve ≥5 mg/mL.
  • Multiple package sizes available for small-scale and bulk needs.