Learn More
Abcam alpha-Methyl 5-hydro x ytryptamine maleate, 5-HT2A/2C receptor agonist, 100MG
Catalog No. 502707496
Encompass_Preferred
MW 306.32 g/mol, Purity >99%. 5-HT₂A/₂C receptor agonist (Ki values are 4.6 and 8.3 nM, respectively in human receptors). Potent antinociceptive effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7496 Supplier Abcam Supplier No. AB120555100MG
MW 306.32 g/mol, Purity >99%. 5-HT₂A/₂C receptor agonist (Ki values are 4.6 and 8.3 nM, respectively in human receptors). Potent antinociceptive effects.
The product is subject to the following: Abcam Restricted Use Statement