Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alpha-Methyl-D-Phenylalanine

Catalog No. 690791250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
69-079-1250MG 100 mg
1 options
Warning

Catalog No. 69-079-1250MG

Supplier: Sigma Aldrich 690791250MG

Only null left
Add to cart
Add to cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alpha-Methyl-D-Phenylalanine

Specifications

CAS 944450-48-0
Molecular Formula C42H34O8P2Ru
Linear Formula C42H34O8P2Ru
MDL Number MFCD09753024
Quantity 100 mg
Synonym Diacetato[(R)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]ruthenium(II)
Molecular Weight (g/mol) 829.73 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.