Learn More
Medchemexpress LLC Alpha-methyl-DL-tryptophan | 153-91-3 | MFCD00038416 | 99.9% | 218.26 g/mol | C12H14N2O2 | 1 ML
SDP
Catalog No. 5000282911
Encompass_Preferred
Alpha-methyl-DL-tryptophan is a tryptophan derivative used as a research reagent that blocks the SLC6A14 transporter and influences mTOR-related signaling in cell models. It is provided as a solid and as a ready-to-use 10 mM solution in DMSO for biochemical and cell-based studies.
  • Ready-to-use 10 mM solution in DMSO (1 mL) available.
  • Also available as solid in multiple mass sizes (10 mg-500 mg).
  • High purity (99.92%) suitable for biochemical assays.
  • Identified by CAS number 153-91-3 for unambiguous sourcing.
  • Used to study apoptosis, autophagy, and mTOR pathway modulation in cell-based research.
Catalog No. 50-002-82911 Supplier Medchemexpress LLC Supplier No. HYW14208010MM/1ML
Alpha-methyl-DL-tryptophan is a tryptophan derivative used as a research reagent that blocks the SLC6A14 transporter and influences mTOR-related signaling in cell models. It is provided as a solid and as a ready-to-use 10 mM solution in DMSO for biochemical and cell-based studies.
  • Ready-to-use 10 mM solution in DMSO (1 mL) available.
  • Also available as solid in multiple mass sizes (10 mg-500 mg).
  • High purity (99.92%) suitable for biochemical assays.
  • Identified by CAS number 153-91-3 for unambiguous sourcing.
  • Used to study apoptosis, autophagy, and mTOR pathway modulation in cell-based research.