Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-Alpha-Methylbenzyl Isocyanate

Catalog No. 2205741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-057-41G 1 g
22-057-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 22-057-41G Supplier Sigma Aldrich Supplier No. 2205741G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(+)-Alpha-Methylbenzyl Isocyanate

Specifications

CAS 33375-06-3
Density 1.045 g/mL at 20°C
Boiling Point 55°C to 56°C (2.5 mmHg, lit.)
Molecular Formula C9H9NO
Linear Formula C6H5CH(CH3)NCO
Refractive Index n20/D 1.513
MDL Number MFCD00063015
Quantity 1 g
Synonym (R)-(+)-1-Phenylethyl isocyanate
Molecular Weight (g/mol) 147.17 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.