Learn More
Medchemexpress LLC Alpha-MSH (alpha-melanocyte-stimulating hormone) | 581-05-5 | 99.98% | 1664.88 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0252
Encompass_Preferred
α-MSH (α-Melanocyte-Stimulating Hormone) is an endogenous neuropeptide that acts as a melanocortin receptor 4 (MC4R) agonist. It exhibits anti-inflammatory and antipyretic activities. This peptide is derived from pro-opiomelanocortin (POMC) through a post-translational modification process.
  • Endogenous neuropeptide
  • Acts as a melanocortin receptor 4 (MC4R) agonist
  • Possesses anti-inflammatory activity
  • Exhibits antipyretic activity
  • Post-translational derivative of pro-opiomelanocortin (POMC)
Catalog No. 50-004-26297