Learn More
TARGETMOL CHEMICALS INC Research Chemical 5mg
SDP
Catalog No. 503038155
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. a-MSH, amide Acetate as an endogenous neuropeptide. It is an endogenous melanocortin receptor 4 (MC4R) agonist with anti-inflammatory and antipyretic activities. (alpha)-MSH is a post-translational derivative of pro-opiomelanocortin (POMC)
Catalog No. 50-303-8155 Supplier TARGETMOL CHEMICALS INC Supplier No. TP2212L5MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. a-MSH, amide Acetate as an endogenous neuropeptide. It is an endogenous melanocortin receptor 4 (MC4R) agonist with anti-inflammatory and antipyretic activities. (alpha)-MSH is a post-translational derivative of pro-opiomelanocortin (POMC)