Learn More
CPC Scientific H-Lys-Pro-Val-NH2 • 2 HCl (hydrochloride salt) 100MG
SDP
Catalog No. 502109826
Encompass
SEQUENCE: H-Lys-Pro-Val-NH2 2 HCl (hydrochloride salt)ONE-LETTER SEQUENCE: KPV-NH2 2 HClMOLECULAR FORMULA: C16H31N5O3MOLECULAR WEIGHT:341.46STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [347870-98-8]SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES:SKU(s): MSH-003A, MSH-003B
Catalog No. 50-210-9826 Supplier CPC Scientific Supplier No. MSH003B
SEQUENCE: H-Lys-Pro-Val-NH2 2 HCl (hydrochloride salt)ONE-LETTER SEQUENCE: KPV-NH2 2 HClMOLECULAR FORMULA: C16H31N5O3MOLECULAR WEIGHT:341.46STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [347870-98-8]SYNONYMS: melanotropinsRESEARCH AREA: HormonalREFERENCES:SKU(s): MSH-003A, MSH-003B