Learn More
CPC Scientific Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific MSHS002A
Encompass
ONE-LETTER SEQUENCE: Ac-SYSMEHFRWGKPVMOLECULAR FORMULA: C77H108N20O20S1MOLECULAR WEIGHT:1665.89STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [10466-28-1], SYNONYMS: melanotropins, acetyl-ACTH (1-13), α-MSH (free acid)RESEARCH AREA: Hormonal
Catalog No. 50-210-9837