Learn More
New England Biolabs, Inc. α-N-Acetylgalactosaminidase - 3000 units
SDP
Catalog No. 50811848
Encompass
a-N-Acetylgalactosaminidase is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing a-N-Acetylgalactosamine residues from oligosaccharides and N-glycans attached to proteins.
Catalog No. 50-811-848 Supplier New England Biolabs, Inc. Supplier No. P0734S
a-N-Acetylgalactosaminidase is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing a-N-Acetylgalactosamine residues from oligosaccharides and N-glycans attached to proteins.