Learn More
Medchemexpress LLC Alpha-N-((R)-1,2-dithiolane-3-pentanoyl)-L-glutamyl-L-alanine | 910627-26-8 | 406.52 g/mol | C16H26N2O6S2 | 5 MG
SDP
Catalog No. 5000227035
Encompass_Preferred
CMX-2043 is a synthetic analogue of α-lipoic acid developed for preclinical research. It shows antioxidant activity, modulates insulin receptor kinase and Akt signaling, and has demonstrated protection in ischemia-reperfusion injury models. Supplied as a powder for preparation of stock solutions.
  • Exhibits antioxidant activity and cell survival pathway modulation.
  • Modulates insulin receptor kinase, tyrosine kinase, and Akt phosphorylation.
  • Shown protective effects in ischemia-reperfusion injury animal models.
  • Supplied as a powder suitable for in vitro and in vivo preclinical studies.
  • Typical pack sizes are intended for small-scale research use.
  • Store sealed, away from moisture; powder: -80°C (2 years) or -20°C (1 year).
Catalog No. 50-002-27035 Supplier Medchemexpress LLC Supplier No. HY1191525MG
CMX-2043 is a synthetic analogue of α-lipoic acid developed for preclinical research. It shows antioxidant activity, modulates insulin receptor kinase and Akt signaling, and has demonstrated protection in ischemia-reperfusion injury models. Supplied as a powder for preparation of stock solutions.
  • Exhibits antioxidant activity and cell survival pathway modulation.
  • Modulates insulin receptor kinase, tyrosine kinase, and Akt phosphorylation.
  • Shown protective effects in ischemia-reperfusion injury animal models.
  • Supplied as a powder suitable for in vitro and in vivo preclinical studies.
  • Typical pack sizes are intended for small-scale research use.
  • Store sealed, away from moisture; powder: -80°C (2 years) or -20°C (1 year).