Learn More
Bioworld Naphthaleneacetic Acid (NAA), 50 g
Catalog No. 50255065
Encompass_Preferred
Alpha-naphthalene acetic acid (NAA)Synonym: 1-Naphthylacetic acidAuxin growth regulatorPlant tissue culture tested Appearance: white to yellow-white crystaline powderMelting Point: 129.0-131.5 °CSoluble in 1M NaOH and AcetoneHarmful if swallowed
Catalog No. 50-255-065 Supplier Bioworld Supplier No. 306310162
Alpha-naphthalene acetic acid (NAA)Synonym: 1-Naphthylacetic acidAuxin growth regulatorPlant tissue culture tested Appearance: white to yellow-white crystaline powderMelting Point: 129.0-131.5 °CSoluble in 1M NaOH and AcetoneHarmful if swallowed