Learn More
Medchemexpress LLC Alpha-naphthoflavone | 604-59-1 | MFCD00004985 | 272.30 g·mol⁻¹ | C19H12O2 | 10MG
SDP
Catalog No. 5000235016
Encompass_Preferred
Alpha-naphthoflavone is a synthetic flavonoid research compound that functions as a potent, competitive aromatase inhibitor and a modulator of cytochrome P450-mediated xenobiotic metabolism. It is used in biochemical and pharmacological studies to probe estrogen biosynthesis and enzyme activity. Chemical identifiers: CAS 604-59-1; formula C19H12O2; molecular weight 272.30 g·mol⁻¹. Supplied as a solid for laboratory use.
  • Potent, competitive aromatase inhibitor for in vitro and biochemical assays.
  • Tool to probe cytochrome P450-mediated metabolism and enzyme induction.
  • Solid form suitable for preparing concentrated stock solutions.
  • Chemical identifiers included for compound verification.
  • Purity not specified on available datasheet; request certificate of analysis if required.
Catalog No. 50-002-35016 Supplier Medchemexpress LLC Supplier No. HY12583310MG
Alpha-naphthoflavone is a synthetic flavonoid research compound that functions as a potent, competitive aromatase inhibitor and a modulator of cytochrome P450-mediated xenobiotic metabolism. It is used in biochemical and pharmacological studies to probe estrogen biosynthesis and enzyme activity. Chemical identifiers: CAS 604-59-1; formula C19H12O2; molecular weight 272.30 g·mol⁻¹. Supplied as a solid for laboratory use.
  • Potent, competitive aromatase inhibitor for in vitro and biochemical assays.
  • Tool to probe cytochrome P450-mediated metabolism and enzyme induction.
  • Solid form suitable for preparing concentrated stock solutions.
  • Chemical identifiers included for compound verification.
  • Purity not specified on available datasheet; request certificate of analysis if required.