Learn More
Medchemexpress LLC Alpha-naphthoflavone | 604-59-1 | MFCD00004985 | ≥98.0% | 272.30 g/mol | C19H12O2 | 20MG
SDP
Catalog No. 5000239103
Encompass_Preferred
Alpha-naphthoflavone is an orally active flavonoid used in research as a potent, competitive inhibitor of aromatase and certain cytochrome P450 enzymes. It is intended for laboratory research use; consult the safety data sheet for handling, storage, and hazard information.
Catalog No. 50-002-39103 Supplier Medchemexpress LLC Supplier No. HY12583320MG
Alpha-naphthoflavone is an orally active flavonoid used in research as a potent, competitive inhibitor of aromatase and certain cytochrome P450 enzymes. It is intended for laboratory research use; consult the safety data sheet for handling, storage, and hazard information.