Learn More
AdipoGen alpha-Naphtholbenzein
SDP
Supplier:  AdipoGen CDXN0116G010
Encompass_Preferred
Chemical. CAS 145-50-6. Formula C27H18O2. MW 374.43. alpha-Naphtholbenzein is a dye generally used as an acid-base indicated. The pH-indicator has a visual transition from yellow pH0-8.2 to turquoise pH10. It is often used during sequential injection analysis technique done for acid-base titrations. It also has been used for food storage, determining bacterial growth in packed food, detecting viable cells and detecting enzymes.
Catalog No. 50-256-0408