Learn More
Sigma Aldrich Fine Chemicals Biosciences alpha-Naphthyl Butyrate Es
Catalog No. 502900882
Encompass
This kit is intended for demonstration of nonspecific esterase in a subset of lymphocytes for determining maturation arrest. Kits 90A1 or 91A are intended for monocyte identification.
Catalog No. 50-290-0882 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 181B1KT
This kit is intended for demonstration of nonspecific esterase in a subset of lymphocytes for determining maturation arrest. Kits 90A1 or 91A are intended for monocyte identification.