Learn More
AdipoGen alpha-NETA (25 mg)
SDP
Supplier:  AdipoGen AGCR10151M025
Encompass_Preferred
Chemical. CAS: 31059-54-8. Formula: C16H20INO. MW: 369.2. Potent fluorescent inhibitor of choline acetyltransferase. CMKLR1 antagonist. Inhibits chemerin-stimulated beta-arrestin2 association with CMKLR1, as well as chemerin-triggered CMKLR1+ cell migration. Aldehyde dehydrogenase 1 (ALDH1A1) antagonist.
Catalog No. 50-205-3853