Learn More
Electron Microscopy Sciences Alpha-Numeric Oven Tray
SDP
Catalog No. 5024491
Encompass_Preferred
Made from polypropylene, Each line is spaced 1 cm apart with letters along the sides and numbers across the top and bottom. This tray has ruled lines that aid in the reproducible placement of the specimens in the oven. Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA.
Catalog No. 50-244-91 Supplier Electron Microscopy Sciences Supplier No. 9703501
Made from polypropylene, Each line is spaced 1 cm apart with letters along the sides and numbers across the top and bottom. This tray has ruled lines that aid in the reproducible placement of the specimens in the oven. Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA.