Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Octithiophene, TCI America™
SDP

Catalog No. O0313100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0313100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O0313100MG Supplier TCI America Supplier No. O0313100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 113728-71-5
Molecular Formula C32H18S8
Molecular Weight (g/mol) 658.98
MDL Number MFCD09038539
InChI Key GCMCTPRNKVKGCY-UHFFFAOYSA-N
PubChem CID 11388231
IUPAC Name 5-{[2,2'-bithiophen]-5-yl}-5'-(5'-{[2,2'-bithiophen]-5-yl}-[2,2'-bithiophen]-5-yl)-2,2'-bithiophene
SMILES S1C=CC=C1C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Melting Point 364°C
Quantity 100 mg
Formula Weight 658.98
Physical Form Crystalline Powder
Chemical Name or Material alpha-Octithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.