Learn More
Medchemexpress LLC Alpha-phellandrene | 99-83-2 | 86.7% | 5 G
SDP
Catalog No. 5000234838
Encompass_Preferred
Alpha-phellandrene (CAS 99-83-2) is a cyclic monoterpene research reagent with reported insecticidal activity. It is a colorless to light yellow liquid (density 0.848 g/cm³) with molecular formula C10H16 and molecular weight 136.23 g/mol.
  • High-purity reagent (86.7% typical) suitable for research and analytical applications
  • Available in small laboratory quantities, including 5 G
  • Soluble in DMSO for solution preparation
  • Commonly used in studies of insecticidal activity and essential oil composition
  • Store in a cool, well-ventilated place away from heat sources
Catalog No. 50-002-34838 Supplier Medchemexpress LLC Supplier No. HY1216155G
Alpha-phellandrene (CAS 99-83-2) is a cyclic monoterpene research reagent with reported insecticidal activity. It is a colorless to light yellow liquid (density 0.848 g/cm³) with molecular formula C10H16 and molecular weight 136.23 g/mol.
  • High-purity reagent (86.7% typical) suitable for research and analytical applications
  • Available in small laboratory quantities, including 5 G
  • Soluble in DMSO for solution preparation
  • Commonly used in studies of insecticidal activity and essential oil composition
  • Store in a cool, well-ventilated place away from heat sources