Learn More
Description
- Adsorption and thermal chemistry of 2,5-dihydrofuran on pd (111) has been studied by high-resolution electron energy loss spectroscopy and temperature-programmed desorption
- Free jet millimetre wave spectrum of the 2,5-dihydrofuran-argon molecular complex has been investigated in the frequency range 60-78ghz
Specifications
Specifications
| CAS | 2510-95-4 |
| Melting Point | 85°C to 87°C (lit.) |
| Molecular Formula | C15H11N |
| Linear Formula | C6H5CH=C(C6H5)CN |
| MDL Number | MFCD00001864 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 205.25 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.