Learn More
Medchemexpress LLC Alpha-phthalimidopropiophenone | 19437-20-8 | MFCD00060520 | >98.0% | 279.3 | C17H13NO3 | 1MG
SDP
Catalog No. 5000236266
Encompass_Preferred
α-Phthalimidopropiophenone is an analytical reference standard (CAS 19437-20-8) consisting of a phthalimido-substituted propiophenone used primarily for forensic and research applications. The compound is a cathinone analog that may act as a prodrug; its physiological and toxicological properties are not well characterized. The phthalimido protecting group can increase stability compared with unsubstituted cathinone analogs.
  • Analytical reference standard suitable for forensic and research use.
  • High purity, ≥98%.
  • Molecular weight 279.3 and formula C17H13NO3.
  • Available in small reference sizes (1 mg-10 mg).
  • May act as a prodrug to a cathinone; physiological effects not well characterized.
Catalog No. 50-002-36266 Supplier Medchemexpress LLC Supplier No. HY1260251MG
α-Phthalimidopropiophenone is an analytical reference standard (CAS 19437-20-8) consisting of a phthalimido-substituted propiophenone used primarily for forensic and research applications. The compound is a cathinone analog that may act as a prodrug; its physiological and toxicological properties are not well characterized. The phthalimido protecting group can increase stability compared with unsubstituted cathinone analogs.
  • Analytical reference standard suitable for forensic and research use.
  • High purity, ≥98%.
  • Molecular weight 279.3 and formula C17H13NO3.
  • Available in small reference sizes (1 mg-10 mg).
  • May act as a prodrug to a cathinone; physiological effects not well characterized.