Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich alpha-Pinene

Catalog No. 1475241L
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-752-41L 1 L
50-185-8971 250 mL
14-752-45ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-752-41L Supplier Sigma Aldrich Supplier No. 1475241L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

alpha-Pinene

Specifications

CAS 80-56-8
Density 0.858 g/mL (at 25°C (literature))
Boiling Point 155°C to 156°C (lit.)
Molecular Formula C10H16
Refractive Index n20/D 1.465 (literature)
Linear Formula C10H16
MDL Number MFCD00001339
Quantity 1 L
Synonym alpha-Pinene; (+/-)-2-Pinene; 2,6,6-Trimethylbicyclo[3.1.1 ]hept-2-ene
Molecular Weight (g/mol) 136.23
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.