Learn More
Texwipe Alpha™ Polyester Knit Cleanroom Swab with Flexible Angled Head
SDP
Catalog No. 17377854 Shop All Texwipe Products
Change view
Click to view available options
Tip Style:
Flexible
Catalog No. Tip Style
17-377-854 Flexible
1 options
Catalog No. 17-377-854 Supplier Texwipe Supplier No. TX761PM
Add to Cart
Add to Cart
Cleanroom Swab features 100% polyester knit construction and a flexible 45° angled head for hard-to-reach areas
  • 45 degree flexible angled head
  • Double layer of polyester knit fabric for enhanced absorbency
  • Engineered to resist snagging and abrading, preventing the release of particles and fibers
  • Excellent chemical compatibility with a variety of solutions
  • 100% virgin polypropylene handle ensures no additional contaminants are introduced while offering excellent chemical resistance
  • Cleanroom laundered, providing ultra-low levels of particles, NVRs, and ions
  • Trademarked light-green colored handles with TEXWIPE name embossed on handle
  • Autoclave safe in dry heat and steam
Specifications
Product Type Cleanroom Swab
Autoclavable Not Autoclavable
Sterility Non-sterile
Adhesive Type Not Applicable
Handle Material Polypropylene
Tip Style Flexible
Tip Material Polyester