Learn More
Texwipe Alpha™ Polyester Knit Large with Notched Handle
SDP
Catalog No. 17377853 Shop All Texwipe Products
Change view
Click to view available options
Tip Style:
Broad
Catalog No. Tip Style
17-377-853 Broad
1 options
Catalog No. 17-377-853 Supplier Texwipe Supplier No. TX718
Add to Cart
Add to Cart
Designed for cleaning validation and surface sampling using HPLC/UV-Vis and chromatography techniques
  • All Surface Cleaning Validation swabs are precision-manufactured to exact and consistent tolerances without the use of adhesives for superior performance.
  • Notched break-away handle allows the swab head to be placed into the vial with minimal handling or contamination
  • 100% polypropylene translucent handle has no colorants or titanium dioxide, providing excellent chemical resistance while reducing potential interferences or contamination
  • 100% Low titanium dioxide knit polyester fabric head
  • Designed for organic solvent-based applications
  • Included in the TX3360 cleaning validation kit
  • Lot coded for traceability and quality control
Specifications
Product Type Polyester Series Swab
Autoclavable Not Autoclavable
Sterility Non-sterile
Adhesive Type Not Applicable
Handle Material Polypropylene
Tip Style Broad
Tip Material Polyester