Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha Sarcoglycan Antibody (YA2068) | 10 UL
SDP

Catalog No. 5000444973
Encompass_Preferred

The alpha Sarcoglycan Antibody (YA2068) is a Rabbit-derived, non-conjugated IgG monoclonal antibody designed to target alpha Sarcoglycan. Alpha Sarcoglycan is a component of the sarcoglycan complex, which forms a structural link between the F-actin cytoskeleton and the extracellular matrix.

  • Applications include WB, IHC-P, and IP
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Liquid appearance
  • Rabbit host
  • Reactivity with human, mouse, and rat samples
  • Non-conjugated
  • Affinity purified

Catalog No. 50-004-44973 Supplier Medchemexpress LLC Supplier No. HYP8232310UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The alpha Sarcoglycan Antibody (YA2068) is a Rabbit-derived, non-conjugated IgG monoclonal antibody designed to target alpha Sarcoglycan. Alpha Sarcoglycan is a component of the sarcoglycan complex, which forms a structural link between the F-actin cytoskeleton and the extracellular matrix.

  • Applications include WB, IHC-P, and IP
  • Supplied in 50mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.01% Sodium azide and 0.05% BSA
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Liquid appearance
  • Rabbit host
  • Reactivity with human, mouse, and rat samples
  • Non-conjugated
  • Affinity purified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.