Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha Sarcoglycan Antibody (YA2068) | 100 UL
SDP

Catalog No. 5000444621
Encompass_Preferred

The alpha Sarcoglycan Antibody (YA2068) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed for various research applications. It targets alpha Sarcoglycan, a component of the sarcoglycan complex involved in the structural link between the F-actin cytoskeleton and the extracellular matrix. This antibody is affinity purified and supplied as a liquid solution.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Observed molecular weight: 50 kDa
  • Applications: Western blot, immunohistochemistry-paraffin, immunoprecipitation
  • Storage at -20°C for 1 year

Catalog No. 50-004-44621 Supplier Medchemexpress LLC Supplier No. HYP82323100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The alpha Sarcoglycan Antibody (YA2068) is a rabbit-derived, non-conjugated IgG monoclonal antibody designed for various research applications. It targets alpha Sarcoglycan, a component of the sarcoglycan complex involved in the structural link between the F-actin cytoskeleton and the extracellular matrix. This antibody is affinity purified and supplied as a liquid solution.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Observed molecular weight: 50 kDa
  • Applications: Western blot, immunohistochemistry-paraffin, immunoprecipitation
  • Storage at -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.