Learn More
Alpha Scientific™ SEG·SAFE™ Segment Processor
Catalog No. 15059350
Change view
Click to view available options
Quantity:
250 Pk.
Catalog No. Quantity
15-059350 250 Pk.
1 options
Catalog No. 15-059350 Supplier Alpha Scientific™ Supplier No. 201
Add to Cart
Edge
Add to Cart
Safe and efficient blood bank device used to process segments for blood grouping/typing or crossmatching
  • The device is comprised of a protected stainless-steel cannula for puncturing the segment, a soft plastic body for holding the cell suspension and a spout for dispensing calibrated drops
  • Can be spun in any standard centrifuge for 12 × 75mm sized test tubes
  • SEG•SAFE is a nonsterile, single-use device and should not be cleaned and reused
  • Dispose of product in an appropriate biohazard container
Hemolysis may occur when using this product; the possibility of hemolysis increases if the blood in the segment is clotted or if the segment is excessively squeezed during piercing.
Specifications
Product Type Segment Processor
Material Plastic
Quantity 250 Pk.
For Use With (Application) Safe and efficient Blood bank device used to process segments for Blood grouping/typing or crossmatching