Learn More
CPC Scientific MCA-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys(DNP)-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108740
Encompass
ONE-LETTER SEQUENCE: Mca-HQKLVFFA-K(DNP)-NH2MOLECULAR FORMULA: C73H94N16O19MOLECULAR WEIGHT:1498.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: M. Farzan, et al., Proc. Natl. Acad. Sci. USA 97, 9712 (2000); T. Kowalewski, and D.M. Holtzman, Proc. Natl. Acad. Sci. USA 96, 3688 (1999); U.S. Ladror, et al., J. Protein. Chem. 13, 357 (1994)
Catalog No. 50-210-8740 Supplier CPC Scientific Supplier No. AMYD112A
ONE-LETTER SEQUENCE: Mca-HQKLVFFA-K(DNP)-NH2MOLECULAR FORMULA: C73H94N16O19MOLECULAR WEIGHT:1498.7STORAGE CONDITIONS: -20 5CRESEARCH AREA: Alzheimer's DiseaseREFERENCES: M. Farzan, et al., Proc. Natl. Acad. Sci. USA 97, 9712 (2000); T. Kowalewski, and D.M. Holtzman, Proc. Natl. Acad. Sci. USA 96, 3688 (1999); U.S. Ladror, et al., J. Protein. Chem. 13, 357 (1994)