Learn More
ACROBiosystems ACROBiosystems Human Alpha-Synuclein Protein, His Tag
Catalog No. 501956412
Encompass_Preferred
Recombinant Protein;200UG;Human Alpha-Synuclein, His Tag (ALN-H52H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.3 kDa.
Catalog No. 50-195-6412 Supplier ACROBiosystems Supplier No. ALNH52H8200UG
Recombinant Protein;200UG;Human Alpha-Synuclein, His Tag (ALN-H52H8) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.3 kDa.