Learn More
Enzo Life Sciences alpha-Synuclein (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201265C100
Encompass_Preferred
Concentration: 1mg/ml. Endotoxin: <1.0EU/µg of protein (LAL method). Purity: ≥95% (SDS-PAGE). MW: ~14.5kDa (MW in SDS-PAGE will be shifted up). Formulation: Liquid. In 20mM TRIS-HCl, pH 7.5, containing 0.1M sodium chloride and 1mM magnesium dichloride. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-200-7961