Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-terpinene | 99-86-5 | MFCD00001534 | 95.0% | 136.23 g·mol⁻¹ | C10H16 | 100 MG
SDP

Catalog No. 5000302038
Encompass_Preferred

α-Terpinene is a research-grade monoterpene (CAS 99-86-5) naturally present in many essential oils. Supplied for laboratory research and analytical applications, it is used in studies of antimicrobial, antifungal, and antioxidant activity. This material is for research use only and not for human consumption.

  • Purity: 95.0% (as supplied)
  • Molecular formula: C10H16
  • Molecular weight: 136.23 g·mol⁻¹
  • Available sizes: 10 mM (1 mL), 100 mg, 500 mg, 1 g
  • Intended for research and analytical use
  • Typical appearance: colorless to light yellow liquid

Catalog No. 50-003-02038 Supplier Medchemexpress LLC Supplier No. HYW020182100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Terpinene is a research-grade monoterpene (CAS 99-86-5) naturally present in many essential oils. Supplied for laboratory research and analytical applications, it is used in studies of antimicrobial, antifungal, and antioxidant activity. This material is for research use only and not for human consumption.

  • Purity: 95.0% (as supplied)
  • Molecular formula: C10H16
  • Molecular weight: 136.23 g·mol⁻¹
  • Available sizes: 10 mM (1 mL), 100 mg, 500 mg, 1 g
  • Intended for research and analytical use
  • Typical appearance: colorless to light yellow liquid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.