Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Alpha-Terpineol

Catalog No. 43262850ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-262-850ML 50 mL
43-262-8250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-262-850ML Supplier Sigma Aldrich Supplier No. 43262850ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Alpha-Terpineol

Specifications

CAS 98-55-5
Melting Point 31°C to 35°C
Density 0.93 g/mL at 25°C
Boiling Point 217°C to 218°C
Molecular Formula C10H18O
Linear Formula C10H18O
Refractive Index n20/D 1.482
MDL Number MFCD00001557
Quantity 50 mL
Synonym alpha-Terpineol
Molecular Weight (g/mol) 154.25 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.