Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-terpineol | 98-55-5 | 5 G
SDP

Catalog No. 5000420078
Encompass_Preferred

Alpha-terpineol is an orally active compound found in Eucalyptus globulus Labill. It exhibits strong antimicrobial activity against periodontopathic and cariogenic bacteria, as well as antifungal activity against T. mentagrophytes, which may lead to irreversible cellular disruption. Additionally, it possesses antineuropathic and anti-inflammatory properties, making it suitable for research related to diarrhea, neuropathic pain, infections, and inflammation.

  • Exhibits strong antimicrobial activity against periodontopathic and cariogenic bacteria
  • Shows antifungal activity against T. mentagrophytes
  • Demonstrates antineuropathic and anti-inflammatory properties
  • Purity of 99.74%
  • Molecular weight of 154.25
  • Appears as a solid below 31°C and a liquid above 35°C
  • Colorless to off-white appearance

Catalog No. 50-004-20078 Supplier Medchemexpress LLC Supplier No. HYN51425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-terpineol is an orally active compound found in Eucalyptus globulus Labill. It exhibits strong antimicrobial activity against periodontopathic and cariogenic bacteria, as well as antifungal activity against T. mentagrophytes, which may lead to irreversible cellular disruption. Additionally, it possesses antineuropathic and anti-inflammatory properties, making it suitable for research related to diarrhea, neuropathic pain, infections, and inflammation.

  • Exhibits strong antimicrobial activity against periodontopathic and cariogenic bacteria
  • Shows antifungal activity against T. mentagrophytes
  • Demonstrates antineuropathic and anti-inflammatory properties
  • Purity of 99.74%
  • Molecular weight of 154.25
  • Appears as a solid below 31°C and a liquid above 35°C
  • Colorless to off-white appearance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.