Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Alpha-terpinyl acetate | 80-26-2 | 5 G
SDP

Catalog No. 5000418126
Encompass_Preferred

Alpha-Terpinyl acetate is a monoterpene ester derived from Laurus nobilis L. essential oil. It acts as a competitive P450 2B6 substrate, binding to the active site with a Kd value of 5.4 μM. It also inhibits bupropion hydroxylation activity of P450 2B6 with an IC50 value of 10.4 μM.

  • Purity: 93.88%
  • Appearance: liquid, colorless to light yellow
  • Molecular weight: 196.29
  • Formula: C12H20O2
  • Classified as: terpenoids, other monoterpenes
  • Derived from: Laurus nobilis L. plants
  • Storage (pure): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility: DMSO: 100 mg/mL

Catalog No. 50-004-18126 Supplier Medchemexpress LLC Supplier No. HYN71365G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alpha-Terpinyl acetate is a monoterpene ester derived from Laurus nobilis L. essential oil. It acts as a competitive P450 2B6 substrate, binding to the active site with a Kd value of 5.4 μM. It also inhibits bupropion hydroxylation activity of P450 2B6 with an IC50 value of 10.4 μM.

  • Purity: 93.88%
  • Appearance: liquid, colorless to light yellow
  • Molecular weight: 196.29
  • Formula: C12H20O2
  • Classified as: terpenoids, other monoterpenes
  • Derived from: Laurus nobilis L. plants
  • Storage (pure): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month
  • Solubility: DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.