Learn More
TARGETMOL CHEMICALS INC ALPHA-THUJONE 100MG
SDP
Catalog No. 502576257
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (alpha)-Thujone is an inhibitor of ACh with an IC50 value of 24.7uM. purity: 98%
Catalog No. 50-257-6257 Supplier TARGETMOL CHEMICALS INC Supplier No. T8193100MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (alpha)-Thujone is an inhibitor of ACh with an IC50 value of 24.7uM. purity: 98%